Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028402.2 | 1 | 76 | |||
GenBank mRNA | AK023759.1 | - | 1 | 76 | ||
AU140233.1 | - | 1 | 76 | |||
BC009174.2 | - | 1 | 51 | 3844322 |