Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003529.3 | 3 | 729 | |||
NR_047539.1 | 3 | 729 | ||||
GenBank mRNA | BI860238.1 | - | 4 | 544 | ||
DQ485453.1 | - | 3 | 728 | |||
DQ485454.1 | - | 3 | 728 |
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF109294.1 | - | 6 | 824 |