Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MGAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002410.4 | NP_002401.1 | 16 | 3338 | ||
XM_005263668.4 | XP_005263725.1 | 17 | 3703 | |||
XM_005263669.4 | XP_005263726.1 | 17 | 3700 | |||
XM_005263670.3 | XP_005263727.1 | 17 | 3356 | |||
XM_006712534.3 | XP_006712597.1 | 18 | 3552 | |||
XM_011511199.2 | XP_011509501.1 | 18 | 3987 | |||
XM_011511200.1 | XP_011509502.1 | 17 | 3352 | |||
XM_011511201.2 | XP_011509503.1 | 17 | 3456 | |||
XM_011511202.1 | XP_011509504.1 | 17 | 3353 | |||
XM_017004147.1 | XP_016859636.1 | 18 | 3827 | |||
XM_017004148.1 | XP_016859637.1 | 17 | 3664 | |||
GenBank mRNA | BU191961.1 | - | 1 | 521 | ||
BX647087.1 | - | 1 | 496 |