Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002766.2 | 3 | 414 | |||
NR_003530.2 | 3 | 414 | ||||
NR_003531.3 | 3 | 390 | ||||
NR_033358.1 | 3 | 414 | ||||
NR_033359.1 | 3 | 414 | ||||
NR_033360.1 | 3 | 414 | ||||
NR_046464.1 | 3 | 414 | ||||
NR_046465.2 | 3 | 414 | ||||
NR_046466.1 | 3 | 414 | ||||
NR_046467.1 | 3 | 414 | ||||
NR_046469.1 | 3 | 414 | ||||
NR_046470.2 | 3 | 414 | ||||
NR_046471.1 | 3 | 414 | ||||
NR_046472.1 | 3 | 414 | ||||
NR_046473.1 | 3 | 414 | ||||
GenBank mRNA | AB032607.1 | - | 3 | 403 | ||
AF052114.1 | - | 1 | 156 | |||
AY314975.1 | - | LIT3321 MEG3 |
3 | 390 | ||
BC023543.2 | - | 3 | 389 | |||
DB449068.1 | - | 3 | 413 | |||
GQ183494.1 | - | 3 | 406 | |||
GQ183495.1 | - | 3 | 406 | |||
GQ183496.1 | - | 3 | 406 | |||
GQ183497.1 | - | 3 | 406 | |||
GQ183498.1 | - | 3 | 406 | |||
GQ183499.1 | - | 3 | 406 | |||
GQ183500.1 | - | 3 | 406 | |||
GQ183501.1 | - | 3 | 406 | |||
GQ183502.1 | - | 3 | 406 | |||
GQ183503.1 | - | 3 | 406 |