Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vax2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002873.2 | 2 | 432 | |||
NR_075094.1 | 2 | 306 | ||||
NR_075095.1 | 2 | 432 | ||||
GenBank mRNA | AK044313.1 | - | LIT3405 Vax2OS |
2 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642242.1 | - | 2 | 307 | Chr. 6 - 83703461 - 83710806 [-] on Build GRCm38 |