Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003718.2 | 7 | 6916 | |||
NR_033657.1 | 7 | 6960 | ||||
GenBank mRNA | AB300594.1 | - | 7 | 6903 | ||
AK028326.1 | - | LIT3370 RNCR2 |
1 | 1255 |