Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610100L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033490.1 | 2 | 290 | |||
GenBank mRNA | AK011787.1 | - | LIT3367 2610100L16 |
2 | 265 | |
AK163433.1 | - | 2 | 163 | |||
BE647934.1 | - | 2 | 290 |
Gene Symbol: A930005K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080704.1 | - | 2 | 587 |