Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snord116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF241256.1 | - | LIT1355 PWCR1 |
1904 |
Gene Symbol: Ipw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045353.1 | - | 1 | 856 |