Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110636.1 | 4 | 6468 | |||
NR_110638.1 | 4 | 2570 | ||||
NR_110640.1 | 5 | 1901 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DQ099683.1 | - | 5 | 1862 | Chr. 9 - 72706244 - 72723120 [-] on Build Rnor_6.0 |