Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_109974.1 | 7 | 1330 | |||
GenBank mRNA | AJ412032.1 | - | LIT1916 BCMS |
7 | 1478 |