Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNHG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002909.2 | 3 | 1449 | |||
NR_036473.1 | 4 | 1557 | ||||
GenBank mRNA | AJ006835.1 | - | LIT1315 U17HG |
3 | 1349 | |
AK311577.1 | - | 1 | 1083 |