Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015342.2 | 3 | 3735 | |||
NR_132312.1 | 4 | 3900 | ||||
NR_132313.1 | 3 | 3600 | ||||
GenBank mRNA | AF103907.1 | - | LIT1552 DD3 |
4 | 3901 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF103908.3 | - | 1 | 5413 | Chr. 9 - 76782136 - 76787569 [+] on Build GRCh38 |