Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK126519.1 | - | 1 | 365 | ||
AK310185.1 | - | 4 | 553 | |||
AY927608.1 | - | LIT3598 AY927608 |
2 | 380 | ||
AY927609.1 | - | 4 | 578 |