Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK057701.1 | - | LIT1007 Ftx |
4 | 692 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB100925.1 | - | 4 | 621 | Chr. X - 74280126 - 74292529 [-] on Build GRCh38 |