Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033796.2 | 1 | 273 | |||
GenBank mRNA | U72518.1 | - | LIT2102 destrin-2 pseudogene |
1 | 273 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN483017.1 | - | 1 | 350 | Chr. 12 - 6884605 - 6885242 [+] on Build GRCh38 |