Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB046220.1 | - | 1 | 369 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI762331.3 | - | 1 | 351 | Chr. 5 - 140374689 - 140375143 [-] on Build GRCh38 |