Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORD116@ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF241255.1 | - | LIT1354 PWCR1 |
1 | 562 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF194765.1 | - | 1 | 13 | Chr. 15 - 25088210 - 25088722 [+] on Build GRCh38 |