Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273635.1 | XP_005273692.1 | 11 | 14680 | ||
XM_017013823.1 | XP_016869312.1 | 11 | 14715 | |||
XM_017013824.1 | XP_016869313.1 | 11 | 14532 | |||
GenBank mRNA | BX648950.1 | - | 1 | 5395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ722936.1 | - | 1 | 82 | Chr. 8 - 29064080 - 29064921 [+] on Build GRCh38 |