Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM198B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036476.1 | 5 | 2745 | |||
NR_036477.1 | 4 | 2652 | ||||
NR_036478.1 | 6 | 2401 | ||||
NR_036479.1 | 5 | 2257 | ||||
GenBank mRNA | AF370398.1 | - | 2 | 1423 | ||
AK057179.1 | - | 4 | 1573 | |||
AK094261.1 | - | 1 | 1426 | |||
AK095340.1 | - | 5 | 2120 | |||
BC072431.1 | - | 5 | 1289 | |||
BX648969.1 | - | 3 | 1494 |