Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010852.3 | NP_001010852.2 | 6 | 9881 | ||
XM_017010266.1 | XP_016865755.1 | 6 | 11583 | |||
GenBank mRNA | AK055705.1 | - | 1 | 1025 |