Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005278385.4 | XP_005278442.1 | 6 | 2759 | ||
GenBank mRNA | BC034279.1 | - | 1 | 908 |