Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038374.1 | 3 | 3903 | |||
NR_038375.1 | 2 | 3801 | ||||
NR_038376.1 | 2 | 3680 | ||||
GenBank mRNA | AL080132.1 | - | 1 | 2623 | ||
BC028422.1 | - | 2 | 3724 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB524548.1 | - | 1 | 370 | Chr. 10 - 28789313 - 28789791 [-] on Build GRCh38 |