Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027207.1 | XP_016882696.1 | 4 | 3854 | ||
XM_017027209.1 | XP_016882698.1 | 4 | 3587 | |||
XM_017027210.1 | XP_016882699.1 | 3 | 3378 | |||
XM_017027211.1 | XP_016882700.1 | 4 | 3521 | |||
XM_017027213.1 | XP_016882702.1 | 4 | 3629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK097763.1 | - | 1 | 1484 | Chr. 19 - 21839597 - 21841647 [-] on Build GRCh38 |