Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027207.1 | XP_016882696.1 | 4 | 3040 | ||
XM_017027209.1 | XP_016882698.1 | 4 | 2773 | |||
XM_017027210.1 | XP_016882699.1 | 3 | 2564 | |||
XM_017027211.1 | XP_016882700.1 | 4 | 2707 | |||
XM_017027213.1 | XP_016882702.1 | 4 | 2815 |