Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257570.3 | XP_005257627.1 | 21 | 5376 | ||
XM_011525110.2 | XP_011523412.1 | 19 | 5075 | |||
XM_011525111.2 | XP_011523413.1 | 19 | 5037 | |||
XM_017024936.1 | XP_016880425.1 | 20 | 5449 | |||
XM_017024937.1 | XP_016880426.1 | 20 | 7125 | |||
XM_017024938.1 | XP_016880427.1 | 20 | 5181 | |||
XM_017024939.1 | XP_016880428.1 | 20 | 5452 | |||
GenBank mRNA | BC040714.1 | - | 1 | 1053 |