Search Thermo Fisher Scientific
Gene Symbol: PTPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002830.3 | NP_002821.1 | 27 | 11331 | ||
GenBank mRNA | AK025546.1 | - | 1 | 1766 | ||
BC037838.1 | - | 1 | 2144 |