Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015134.3 | NP_055949.2 | 23 | 9446 | ||
NM_201274.3 | NP_958431.2 | 24 | 9509 | |||
XM_005256563.4 | XP_005256620.1 | 24 | 13558 | |||
XM_005256564.4 | XP_005256621.3 | 22 | 9637 | |||
XM_011523761.2 | XP_011522063.1 | 24 | 13929 | |||
XM_011523762.2 | XP_011522064.1 | 25 | 13975 | |||
XM_011523763.2 | XP_011522065.1 | 23 | 13798 | |||
XM_011523764.2 | XP_011522066.1 | 23 | 13792 | |||
XM_011523765.2 | XP_011522067.1 | 24 | 13855 | |||
XM_011523766.2 | XP_011522068.2 | 23 | 10105 | |||
XM_011523767.2 | XP_011522069.2 | 24 | 10168 | |||
XM_017024393.1 | XP_016879882.1 | 23 | 10048 | |||
GenBank mRNA | AK091337.1 | - | 1 | 749 |