Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096766.1 | - | 1 | 787 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD640803.1 | - | 2 | 190 | Chr. 7 - 38303893 - 38305258 [-] on Build GRCh38 |