Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100289518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026856.1 | - | 1 | 218 |
Gene Symbol: EED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545330.1 | XP_011543632.1 | 13 | 2070 | ||
XM_017018512.1 | XP_016874001.1 | 12 | 1995 | |||
XM_017018513.1 | XP_016874002.1 | 10 | 1755 |