Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036442.1 | 4 | 902 | |||
GenBank mRNA | BC036550.1 | - | 2 | 2875 | ||
BC070105.1 | - | 2 | 2373 | |||
BC098409.1 | - | 2 | 2373 |