Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019912.1 | XP_016875401.1 | 1 | 1393 | ||
GenBank mRNA | BC011028.1 | - | 1 | 634 |