Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: API5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006718359.3 | XP_006718422.1 | 14 | 2798 | ||
XM_017018464.1 | XP_016873953.1 | 15 | 2855 | |||
GenBank mRNA | AK021649.1 | - | 1 | 707 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU118497.1 | - | 1 | 707 | Chr. 11 - 43339677 - 43340535 [+] on Build GRCh38 |