Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303030.1 | NP_001289959.1 | 1 | 261 | ||
NM_001303031.1 | NP_001289960.1 | 1 | 261 | |||
XM_005270975.4 | XP_005271032.1 | 1 | 1116 | |||
XM_005270981.4 | XP_005271038.1 | 1 | 1116 | |||
XM_005270982.1 | XP_005271039.1 | 1 | 195 | |||
XM_011541637.1 | XP_011539939.1 | 3 | 534 | |||
XM_011541638.1 | XP_011539940.1 | 2 | 411 | |||
XM_011541639.1 | XP_011539941.1 | 2 | 411 | |||
XM_011541646.2 | XP_011539948.1 | 1 | 1116 | |||
XM_017001544.1 | XP_016857033.1 | 2 | 503 | |||
XM_017001546.1 | XP_016857035.1 | 1 | 1115 | |||
XM_017001547.1 | XP_016857036.1 | 1 | 195 | |||
XM_017001548.1 | XP_016857037.1 | 3 | 534 | |||
XM_017001549.1 | XP_016857038.1 | 2 | 411 | |||
XM_017001550.1 | XP_016857039.1 | 2 | 411 | |||
GenBank mRNA | AK056029.1 | - | 1 | 2007 | ||
AK299377.1 | - | 1 | 160 | |||
AK316093.1 | - | 1 | 114 | |||
BC063843.1 | - | 1 | 169 | 6501461 | ||
DC314995.1 | - | 1 | 262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX644786.1 | - | 1 | 380 | Chr. 1 - 37689953 - 37690708 [-] on Build GRCh38 |