Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 4642 | ||
XM_005266733.1 | XP_005266790.1 | 7 | 4563 | |||
XM_006722518.1 | XP_006722581.1 | 6 | 1152 | |||
XM_017025913.1 | XP_016881402.1 | 9 | 5396 | |||
XM_017025914.1 | XP_016881403.1 | 8 | 4791 | |||
GenBank mRNA | AF131784.1 | - | 1 | 209 |