Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004676.1 | XP_016860165.1 | 9 | 1899 | ||
XM_017004682.1 | XP_016860171.1 | 9 | 1882 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC035835.1 | - | 2 | 970 | Chr. 2 - 189746662 - 189748981 [-] on Build GRCh38 |