Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532987.2 | XP_011531289.1 | 17 | 7455 | ||
XM_017004503.1 | XP_016859992.1 | 16 | 7322 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR603904.1 | - | 1 | 405 | Chr. 2 - 37099241 - 37102135 [-] on Build GRCh38 |