Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008945.1 | XP_016864434.1 | 18 | 2397 | ||
XM_017008946.1 | XP_016864435.1 | 18 | 2363 | |||
XM_017008947.1 | XP_016864436.1 | 19 | 2866 | |||
XM_017008948.1 | XP_016864437.1 | 17 | 2658 | |||
XM_017008949.1 | XP_016864438.1 | 17 | 2205 | |||
GenBank mRNA | AK097213.1 | - | 1 | 1910 |