Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC106699570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133569.1 | 2 | 1873 | |||
GenBank mRNA | AL080152.1 | - | 2 | 1873 |