Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_109974.1 | 6 | 828 | |||
GenBank mRNA | AJ412026.1 | - | LIT1926 BCMS |
4 | 681 | |
AJ412028.1 | - | LIT1924 BCMS |
4 | 361 | ||
AJ412032.1 | - | LIT1916 BCMS |
6 | 976 | ||
AJ412035.1 | - | LIT1910 BCMS |
2 | 156 | ||
AJ412037.1 | - | LIT1908 BCMS |
3 | 294 | ||
AJ412043.1 | - | LIT1902 BCMS |
5 | 579 | ||
AJ412055.1 | - | LIT1885 BCMS |
6 | 826 | ||
AJ412056.1 | - | LIT1884 BCMS |
6 | 892 | ||
AJ412059.1 | - | LIT1874 BCMS |
7 | 939 | ||
AJ412060.1 | - | LIT1873 BCMS |
5 | 1925 | ||
AJ412061.1 | - | LIT1872 BCMS |
6 | 828 | ||
AJ412062.1 | - | LIT1871 BCMS |
6 | 828 | ||
AJ412063.1 | - | LIT1870 BCMS |
3 | 275 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AJ412015.3 | - | 1 | 43 | Chr. 13 - 50527866 - 50528064 [+] on Build GRCh38 |