Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321265.1 | NP_001308194.1 | 12 | 7342 | ||
NM_001321266.1 | NP_001308195.1 | 11 | 7264 | |||
NM_001321267.1 | NP_001308196.1 | 14 | 7565 | |||
NM_138799.3 | NP_620154.2 | 13 | 7488 | |||
NR_135598.1 | 14 | 7540 | ||||
NR_135599.1 | 13 | 7807 | ||||
NR_135600.1 | 13 | 7394 | ||||
NR_135601.1 | 11 | 7183 | ||||
NR_135602.1 | 12 | 7355 | ||||
NR_135603.1 | 14 | 7581 | ||||
NR_135604.1 | 13 | 7504 | ||||
NR_135605.1 | 12 | 7316 | ||||
NR_135606.1 | 13 | 7407 | ||||
GenBank mRNA | AF052107.1 | - | 1 | 1357 |