Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101060542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110764.1 | 3 | 1184 | |||
GenBank mRNA | CR936624.1 | - | 3 | 1197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000582251.1 | - | 3 | 1184 | Chr. 18 - 70380140 - 70384591 [-] on Build GRCh38 |