Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101060542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110764.1 | 3 | 701 | |||
GenBank mRNA | CR936624.1 | - | 3 | 701 |