Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521414.1 | XP_011519716.1 | 11 | 2874 | ||
XM_017022044.1 | XP_016877533.1 | 11 | 2870 | |||
GenBank mRNA | AK094290.1 | - | 1 | 1931 | ||
BC114951.1 | - | 12 | 2506 | 40005391 |