Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005253351.3 | XP_005253408.1 | 4 | 6530 | ||
XM_011520628.2 | XP_011518930.1 | 14 | 9596 | |||
XM_011520629.2 | XP_011518931.1 | 15 | 9403 | |||
XM_017019219.1 | XP_016874708.1 | 14 | 9151 | |||
GenBank mRNA | AK091524.1 | - | 1 | 2130 |