Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBOX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122033.1 | 2 | 2021 | |||
GenBank mRNA | AK057338.1 | - | 2 | 2021 |