Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029124.1 | - | 1 | 316 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI824106.1 | - | 1 | 317 | Chr. 10 - 122163593 - 122164377 [-] on Build GRCh38 |