Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518646.2 | XP_011516948.1 | 18 | 4358 | ||
GenBank mRNA | AK094719.1 | - | 1 | 1880 |