Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256063.1 | NP_001242992.1 | 1 | 97 | ||
NM_001843.3 | NP_001834.2 | 1 | 97 | |||
XM_017018821.1 | XP_016874310.1 | 1 | 100 | |||
GenBank mRNA | AK289698.1 | - | 1 | 39 | ||
DQ442054.1 | - | 1 | 97 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA192436.1 | - | 1 | 92 | Chr. 12 - 40692447 - 40922391 [+] on Build GRCh38 |