Gene Symbol: CD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510119.2 | XP_011508421.1 | 6 | 3488 | ||
XM_017002786.1 | XP_016858275.1 | 4 | 3280 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK130822.1 | - | 904 | Chr. - - [-] on Build |