Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199633.1 | NP_001186562.1 | 18 | 2840 | ||
NM_022127.2 | NP_071410.1 | 19 | 2786 | |||
NR_037638.2 | 19 | 3016 | ||||
XM_011518905.2 | XP_011517207.1 | 18 | 2910 | |||
XM_011518906.2 | XP_011517208.1 | 19 | 3006 | |||
XM_011518907.2 | XP_011517209.1 | 17 | 2725 | |||
XM_011518908.2 | XP_011517210.1 | 13 | 2745 | |||
GenBank mRNA | AK022042.1 | - | 1 | 582 |